Product Details

SNP ID
rs2289973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85577559 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGCGGGGGCAGGCTGAAAGTTG[A/G]AGCAAGCAGGAAGTGAACTGAGGGC
Phenotype
MIM: 607029 MIM: 603177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VAMP5 PubMed Links

Gene Details

Gene
VAMP5
Gene Name
vesicle associated membrane protein 5
There are no transcripts associated with this gene.

Gene
VAMP8
Gene Name
vesicle associated membrane protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003761.4 25 UTR 5 NP_003752.2
XM_017005170.1 25 UTR 5 XP_016860659.1

View Full Product Details