Product Details

SNP ID
rs2277064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134648839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGTTTGGCTGCCGCCTTCTAGC[C/G]GGGCGTTCGCGGCCCCGCCGGCCCG
Phenotype
MIM: 607183
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SEC24A PubMed Links

Gene Details

Gene
SEC24A
Gene Name
SEC24 homolog A, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252231.1 55 UTR 5 NP_001239160.1
NM_021982.2 55 UTR 5 NP_068817.1
XM_006714523.2 55 UTR 5 XP_006714586.1
XM_017008961.1 55 Intron XP_016864450.1
XM_017008962.1 55 Intron XP_016864451.1
XM_017008963.1 55 UTR 5 XP_016864452.1

View Full Product Details