Product Details

SNP ID
rs2277124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43672327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTGGCTCACTCAGCTACCCCA[A/G]CCTTCAGGGAACTCCCCAGGGTACT
Phenotype
MIM: 611981 MIM: 612648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS18A PubMed Links

Gene Details

Gene
MRPS18A
Gene Name
mitochondrial ribosomal protein S18A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193343.1 Intron NP_001180272.1
NM_018135.3 Intron NP_060605.1
XM_006715134.2 Intron XP_006715197.1
Gene
RSPH9
Gene Name
radial spoke head 9 homolog
There are no transcripts associated with this gene.

View Full Product Details