Product Details

SNP ID
rs2969934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57667921 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTTGGGCGCGGCAGCGCGAGCC[G/T]CATGAATGAGACAAACGTGCGCCGC
Phenotype
MIM: 608757 MIM: 614586
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLP1 PubMed Links

Gene Details

Gene
CLP1
Gene Name
cleavage and polyadenylation factor I subunit 1
There are no transcripts associated with this gene.

Gene
ZDHHC5
Gene Name
zinc finger DHHC-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015457.2 86 Intron NP_056272.2
XM_011544899.1 86 Intron XP_011543201.1
XM_011544900.1 86 Intron XP_011543202.1
XM_011544901.1 86 Intron XP_011543203.1
XM_017017498.1 86 UTR 5 XP_016872987.1

View Full Product Details