Product Details

SNP ID
rs2290387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61349743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTCAGCTCCTCAGCAGAAGCTGC[A/G]TGGGCCGCCACTCCCCCTTTCTGCA
Phenotype
MIM: 615844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYB561A3 PubMed Links

Gene Details

Gene
CYB561A3
Gene Name
cytochrome b561 family member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161452.1 1561 UTR 3 NP_001154924.1
NM_001161454.1 1561 UTR 3 NP_001154926.1
NM_001300763.1 1561 Intron NP_001287692.1
NM_153611.4 1561 UTR 3 NP_705839.3
XM_011544821.1 1561 Intron XP_011543123.1
XM_011544823.1 1561 Intron XP_011543125.1
XM_011544824.1 1561 Intron XP_011543126.1
XM_017017345.1 1561 Intron XP_016872834.1
XM_017017346.1 1561 Intron XP_016872835.1
XM_017017347.1 1561 Intron XP_016872836.1
Gene
TKFC
Gene Name
triokinase and FMN cyclase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015533.3 1561 Intron NP_056348.2
XM_011544912.2 1561 Intron XP_011543214.1
XM_017017517.1 1561 Intron XP_016873006.1
XM_017017518.1 1561 Intron XP_016873007.1
XM_017017519.1 1561 Intron XP_016873008.1
XM_017017520.1 1561 Intron XP_016873009.1
XM_017017521.1 1561 Intron XP_016873010.1
XM_017017522.1 1561 Intron XP_016873011.1
XM_017017523.1 1561 Intron XP_016873012.1
XM_017017524.1 1561 Intron XP_016873013.1

View Full Product Details