Product Details

SNP ID
rs2598043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:37850979 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGAAAGTCAGAAAAGTGTAATA[A/G]TGGACACATCTATTGTTCTGTTTCC
Phenotype
MIM: 607421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NME8 PubMed Links

Gene Details

Gene
NME8
Gene Name
NME/NM23 family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016616.4 Intron NP_057700.3

View Full Product Details