Product Details

SNP ID
rs2708959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112912838 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATGGTCTGGCCCCTTGGTGCCT[A/T]TCTGGTTGCAGCGAGCATTTCAAAC
Phenotype
MIM: 605510
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IL37 PubMed Links

Gene Details

Gene
IL37
Gene Name
interleukin 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014439.3 Intron NP_055254.2
NM_173202.1 Intron NP_775294.1
NM_173203.1 Intron NP_775295.1
NM_173204.1 Intron NP_775296.1
NM_173205.1 Intron NP_775297.1
XM_011510963.2 Intron XP_011509265.1
XM_011510964.2 Intron XP_011509266.1

View Full Product Details