Product Details

SNP ID
rs2710889
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:1023573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCGTGGACTCAGGATAGGCTGTC[A/G]GGAGCTGAGGAGGGGCTGGAGAAGG
Phenotype
MIM: 103320 MIM: 147571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGRN PubMed Links

Gene Details

Gene
AGRN
Gene Name
agrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305275.1 Intron NP_001292204.1
NM_198576.3 Intron NP_940978.2
XM_005244749.3 Intron XP_005244806.1
XM_011541429.2 Intron XP_011539731.1
Gene
ISG15
Gene Name
ISG15 ubiquitin-like modifier
There are no transcripts associated with this gene.

View Full Product Details