Product Details

SNP ID
rs2721358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:120991463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGTCATTTCTTGTTAGTTTCAA[C/T]ATTGAGAAAGGCAATTTCTTAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPED1 PubMed Links

Gene Details

Gene
CPED1
Gene Name
cadherin like and PC-esterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105533.1 Intron NP_001099003.1
NM_024913.4 Intron NP_079189.4
XM_017012649.1 Intron XP_016868138.1

View Full Product Details