Product Details

SNP ID
rs2261295
Assay Type
Validated
NCBI dbSNP Submissions
64
Location
Chr.1:157833189 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CCTTATGAACTCCATCTGTAGGACT[T/C]ACCTGAGCAGATGACAGCCACATCT
Phenotype
MIM: 602592
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CD5L PubMed Links

Gene Details

Gene
CD5L
Gene Name
CD5 molecule like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005894.2 Intron NP_005885.1
XM_005245602.1 Intron XP_005245659.1
XM_017002806.1 Intron XP_016858295.1

View Full Product Details