Product Details

SNP ID
rs2784867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:163419497 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGTGAAATAATCTGTTCTTTAG[A/G]AACTAAGATATTGCTTTTGTACACA
Phenotype
MIM: 615930 MIM: 609590
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAHM PubMed Links

Gene Details

Gene
CAHM
Gene Name
colon adenocarcinoma hypermethylated (non-protein coding)
There are no transcripts associated with this gene.

Gene
QKI
Gene Name
QKI, KH domain containing RNA binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301085.1 Intron NP_001288014.1
NM_006775.2 Intron NP_006766.1
NM_206853.2 Intron NP_996735.1
NM_206854.2 Intron NP_996736.1
NM_206855.2 Intron NP_996737.1
XM_011536258.2 Intron XP_011534560.1
XM_011536259.2 Intron XP_011534561.1
XM_011536260.2 Intron XP_011534562.1
XM_011536261.1 Intron XP_011534563.1
XM_017011502.1 Intron XP_016866991.1
XM_017011503.1 Intron XP_016866992.1
XM_017011504.1 Intron XP_016866993.1

View Full Product Details