Product Details

SNP ID
rs2233308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151033490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGGACACCCACCTGCCACCAC[A/G]GTGCTGTTTAGTTCAGAAGGATGCA
Phenotype
MIM: 138321 MIM: 607714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPX3 PubMed Links

Gene Details

Gene
GPX3
Gene Name
glutathione peroxidase 3
There are no transcripts associated with this gene.

Gene
TNIP1
Gene Name
TNFAIP3 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252385.1 Intron NP_001239314.1
NM_001252386.1 Intron NP_001239315.1
NM_001252390.1 Intron NP_001239319.1
NM_001252391.1 Intron NP_001239320.1
NM_001252392.1 Intron NP_001239321.1
NM_001252393.1 Intron NP_001239322.1
NM_001258454.1 Intron NP_001245383.1
NM_001258455.1 Intron NP_001245384.1
NM_001258456.1 Intron NP_001245385.1
NM_006058.4 Intron NP_006049.3
XM_005268355.1 Intron XP_005268412.1
XM_006714751.1 Intron XP_006714814.1
XM_006714752.2 Intron XP_006714815.1
XM_017008945.1 Intron XP_016864434.1
XM_017008946.1 Intron XP_016864435.1
XM_017008947.1 Intron XP_016864436.1
XM_017008948.1 Intron XP_016864437.1
XM_017008949.1 Intron XP_016864438.1
XM_017008950.1 Intron XP_016864439.1
XM_017008951.1 Intron XP_016864440.1

View Full Product Details