Product Details
- SNP ID
-
rs2233623
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:14979061 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGGCAAACTAAAGGAGTTTCTCA[A/C]ATACAAACACTTAAACCTCTCCTTC
- Phenotype
-
MIM: 601190
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PDE6H
PubMed Links
Gene Details
- Gene
- PDE6H
- Gene Name
- phosphodiesterase 6H
View Full Product Details