Product Details
- SNP ID
-
rs2233763
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8188982 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACACACAGGCCTCCCCTCAGTGGG[C/T]ACTTCCCACCAGGACTCCCTGAAGA
- Phenotype
-
MIM: 616599
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BORCS6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115524948] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BORCS6
- Gene Name
- BLOC-1 related complex subunit 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017622.2 |
1265 |
UTR 3 |
|
|
NP_060092.2 |
- Gene
- MIR4521
- Gene Name
- microRNA 4521
There are no transcripts associated with this gene.
View Full Product Details