Product Details

SNP ID
rs2341323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8849634 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATCGCCTGTCTTCTGTCTAAATA[C/T]CATTTCTCTCTCATCTCATTCTCAG
Phenotype
MIM: 607963 MIM: 606154
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBD3L1 PubMed Links

Gene Details

Gene
MBD3L1
Gene Name
methyl-CpG binding domain protein 3 like 1
There are no transcripts associated with this gene.

Gene
MUC16
Gene Name
mucin 16, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024690.2 Intron NP_078966.2
XM_017027486.1 Intron XP_016882975.1
XM_017027487.1 Intron XP_016882976.1
XM_017027488.1 Intron XP_016882977.1
XM_017027489.1 Intron XP_016882978.1
XM_017027490.1 Intron XP_016882979.1
XM_017027491.1 Intron XP_016882980.1
XM_017027492.1 Intron XP_016882981.1
XM_017027493.1 Intron XP_016882982.1
XM_017027494.1 Intron XP_016882983.1
XM_017027495.1 Intron XP_016882984.1
XM_017027496.1 Intron XP_016882985.1
XM_017027497.1 Intron XP_016882986.1
XM_017027498.1 Intron XP_016882987.1
XM_017027499.1 Intron XP_016882988.1
XM_017027500.1 Intron XP_016882989.1
XM_017027501.1 Intron XP_016882990.1

View Full Product Details