Product Details
- SNP ID
-
rs2234235
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:9629183 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGAACTTTTTTGCATTTTGTTTCA[A/G]TTTAGGATTTCCTAAAATTAAGATG
- Phenotype
-
MIM: 604796
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R1
PubMed Links
Gene Details
- Gene
- TAS2R1
- Gene Name
- taste 2 receptor member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019599.2 |
1169 |
Silent Mutation |
CTG,TTG |
L284L |
NP_062545.1 |
View Full Product Details