Product Details

SNP ID
rs2279159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70301695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCGCCGAGGACGCTCCTGCCCC[A/G]GGCTGGAAGGGGCGGAAAACAGCCT
Phenotype
MIM: 616275 MIM: 603542
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM136A PubMed Links

Gene Details

Gene
FAM136A
Gene Name
family with sequence similarity 136 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032822.2 Intron NP_116211.2
XM_005264620.1 Intron XP_005264677.1
Gene
SNRPG
Gene Name
small nuclear ribonucleoprotein polypeptide G
There are no transcripts associated with this gene.

View Full Product Details