Product Details

SNP ID
rs2399431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112561859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCACACGCACCCCTCGCCCTCTG[C/G]GAGCTGTCTGTCCTCGCTTTGCTTC
Phenotype
MIM: 609606
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATG3 PubMed Links

Gene Details

Gene
ATG3
Gene Name
autophagy related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278712.1 104 UTR 5 NP_001265641.1
NM_022488.4 104 UTR 5 NP_071933.2
XM_011513074.1 104 UTR 5 XP_011511376.1
Gene
SLC35A5
Gene Name
solute carrier family 35 member A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017945.3 104 Intron NP_060415.1
XM_011512948.1 104 Intron XP_011511250.1

View Full Product Details