Product Details
- SNP ID
-
rs2236172
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31441509 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGAGGGGTGGGTTAGGAGAGAGGC[A/G]GGGGTAAAGCTGAGGAAGCATTCAA
- Phenotype
-
MIM: 616076
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB123
PubMed Links
Gene Details
- Gene
- DEFB123
- Gene Name
- defensin beta 123
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153324.3 |
|
Intron |
|
|
NP_697019.1 |
View Full Product Details