Product Details
- SNP ID
-
rs2236363
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:52845004 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATTATACATATTCAGGCTTCCATCT[T/G]CAGCATGTCAGCCTGGGTCAGGGCA
- Phenotype
-
MIM: 603580
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PCDH8
PubMed Links
Gene Details
- Gene
- PCDH8
- Gene Name
- protocadherin 8
View Full Product Details