Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033551.2 | 4993 | UTR 3 | NP_001028723.1 | ||
NM_001082968.1 | 4993 | UTR 3 | NP_001076437.1 | ||
NM_001288786.1 | 4993 | UTR 3 | NP_001275715.1 | ||
NM_001288787.1 | 4993 | UTR 3 | NP_001275716.1 | ||
NM_001288788.1 | 4993 | UTR 3 | NP_001275717.1 | ||
NM_001288789.1 | 4993 | UTR 3 | NP_001275718.1 | ||
XM_005256461.2 | 4993 | UTR 3 | XP_005256518.1 | ||
XM_005256462.1 | 4993 | UTR 3 | XP_005256519.1 | ||
XM_005256463.2 | 4993 | UTR 3 | XP_005256520.1 | ||
XM_005256466.1 | 4993 | UTR 3 | XP_005256523.1 | ||
XM_011523662.1 | 4993 | UTR 3 | XP_011521964.1 | ||
XM_017024201.1 | 4993 | UTR 3 | XP_016879690.1 |