Product Details
- SNP ID
-
rs2236600
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112879483 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGCCTGGGACTGCTGTGTGCAGCCT[C/G]CCTGTAACCCTTAAGGTCATGCTCT
- Phenotype
-
MIM: 608956
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC46A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78210382] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC46A2
- Gene Name
- solute carrier family 46 member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033051.3 |
1939 |
UTR 3 |
|
|
NP_149040.3 |
- Gene
- SNX30
- Gene Name
- sorting nexin family member 30
There are no transcripts associated with this gene.
View Full Product Details