Product Details

SNP ID
rs2227634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101127092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGGTATAAAAGGAGGCAGTGGC[C/T]CACAGAGGAGCACAGCTGTGTTTGG
Phenotype
MIM: 173360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINE1 PubMed Links

Gene Details

Gene
SERPINE1
Gene Name
serpin family E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000602.4 6 UTR 5 NP_000593.1
XM_017012260.1 6 UTR 5 XP_016867749.1

View Full Product Details