Product Details

SNP ID
rs2270998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100559173 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTGGGCACTGAGTGCCGCGCAG[C/T]AGCGGGGAAGATGGGGCCCGTGGCC
Phenotype
MIM: 610350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINS1 PubMed Links

Gene Details

Gene
LINS1
Gene Name
lines homolog 1
There are no transcripts associated with this gene.

Gene
LOC102723335
Gene Name
uncharacterized LOC102723335
There are no transcripts associated with this gene.

Gene
PRKXP1
Gene Name
protein kinase, X-linked, pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details