Product Details

SNP ID
rs2271991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126667048 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTGGGGAAAACAGTCCCGCCTCT[C/G]GTTTAACTGTTTGGTATTTCATCAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUP210P1 PubMed Links

Gene Details

Gene
NUP210P1
Gene Name
nucleoporin 210 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details