Product Details

SNP ID
rs2272063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168924416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGATTATTTGCCTTTGGAAAAAA[A/G]TGTCACTTAGCCTAAAGAATCTTAA
Phenotype
MIM: 603201
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCB11 PubMed Links

Gene Details

Gene
ABCB11
Gene Name
ATP binding cassette subfamily B member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003742.2 Intron NP_003733.2
XM_006712817.3 Intron XP_006712880.1
XM_011512077.2 Intron XP_011510379.1
XM_011512078.2 Intron XP_011510380.1
XM_011512080.2 Intron XP_011510382.1
XM_011512081.2 Intron XP_011510383.1
XM_017005165.1 Intron XP_016860654.1
XM_017005166.1 Intron XP_016860655.1
XM_017005167.1 Intron XP_016860656.1

View Full Product Details