Product Details

SNP ID
rs2974910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35336405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTATGTTTACATTTGCCTTCAA[A/C]ACTGCCATCCGCTACAAATTTTTCC
Phenotype
MIM: 194534
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 2112 Intron NP_001295052.1
NM_006965.3 2112 UTR 3 NP_008896.2
XM_005258341.3 2112 UTR 3 XP_005258398.1

View Full Product Details