Product Details

SNP ID
rs2272368
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:124327309 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCACAAAAATAAAGTCACAAGCTCC[A/G]AAGCTCGAGGAGGGGGTTGTCAGAG
Phenotype
MIM: 600848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6880 PubMed Links

Gene Details

Gene
MIR6880
Gene Name
microRNA 6880
There are no transcripts associated with this gene.

Gene
NCOR2
Gene Name
nuclear receptor corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077261.3 Intron NP_001070729.2
NM_001206654.1 Intron NP_001193583.1
NM_006312.5 Intron NP_006303.4

View Full Product Details