Product Details
- SNP ID
-
rs2963698
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:90481083 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATCCCTGGTGGAATTCCAATTAAT[A/G]AAACAGAAAGAAAGATAAAATTTGT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MBLAC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145118946] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MBLAC2
- Gene Name
- metallo-beta-lactamase domain containing 2
There are no transcripts associated with this gene.
- Gene
- POLR3G
- Gene Name
- RNA polymerase III subunit G
View Full Product Details