Product Details

SNP ID
rs2279010
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:77632932 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCACAGCGAGAAGACCTAAGGCCG[A/C]AAGTCGCTGTGCAGCACCCACCCAA
Phenotype
MIM: 604529
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OTP PubMed Links

Gene Details

Gene
OTP
Gene Name
orthopedia homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032109.2 Intron NP_115485.1

View Full Product Details