Product Details

SNP ID
rs2279280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1251598 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTAGGTCCCCCCGGCCAGCCTCC[A/G]TTGCCACATGGGCTATTTTTGTCCA
Phenotype
MIM: 602618
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTBP1 PubMed Links

Gene Details

Gene
CTBP1
Gene Name
C-terminal binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012614.1 Intron NP_001012632.1
NM_001328.2 Intron NP_001319.1
Gene
CTBP1-AS2
Gene Name
CTBP1 antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

View Full Product Details