Product Details

SNP ID
rs2279373
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56994414 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGACCTCCCCGCCACTGGCACAC[C/T]GGGTCTTTTGCTGGGACATCCCCTG
Phenotype
MIM: 605307
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPR182 PubMed Links
Additional Information
For this assay, SNP(s) [rs76016938] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GPR182
Gene Name
G protein-coupled receptor 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007264.3 520 Intron NP_009195.1
XM_011537824.2 520 UTR 5 XP_011536126.1
XM_011537825.2 520 UTR 5 XP_011536127.1
XM_011537827.2 520 UTR 5 XP_011536129.1
Gene
HBCBP
Gene Name
HBcAg-binding protein
There are no transcripts associated with this gene.

Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
There are no transcripts associated with this gene.

View Full Product Details