Product Details

SNP ID
rs2279533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6926734 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGGAGCGGTGCTCACGGGTATC[A/G]TTCTCGTCAGCGGGCGGTGGGGACG
Phenotype
MIM: 605016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2D3 PubMed Links

Gene Details

Gene
OR2D3
Gene Name
olfactory receptor family 2 subfamily D member 3
There are no transcripts associated with this gene.

Gene
ZNF215
Gene Name
zinc finger protein 215
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013250.2 Intron NP_037382.2
XM_006718311.2 Intron XP_006718374.1

View Full Product Details