Product Details

SNP ID
rs2280004
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:45907766 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTCCAAATGAGTTTTGCTATACGA[A/G]GATGCTGATAATAATAGCAGCAGTG
Phenotype
MIM: 157140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPT PubMed Links

Gene Details

Gene
MAPT
Gene Name
microtubule associated protein tau
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123066.3 Intron NP_001116538.2
NM_001123067.3 Intron NP_001116539.1
NM_001203251.1 Intron NP_001190180.1
NM_001203252.1 Intron NP_001190181.1
NM_005910.5 Intron NP_005901.2
NM_016834.4 Intron NP_058518.1
NM_016835.4 Intron NP_058519.3
NM_016841.4 Intron NP_058525.1
XM_005257362.4 Intron XP_005257419.1
XM_005257364.4 Intron XP_005257421.1
XM_005257365.4 Intron XP_005257422.1
XM_005257366.3 Intron XP_005257423.1
XM_005257367.4 Intron XP_005257424.1
XM_005257368.4 Intron XP_005257425.1
XM_005257369.4 Intron XP_005257426.1
XM_005257370.4 Intron XP_005257427.1
XM_005257371.4 Intron XP_005257428.1
Gene
MAPT-IT1
Gene Name
MAPT intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details