Product Details

SNP ID
rs2280089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3669480 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACTGGGTGCTGCCCATCTGGAAG[A/G]AGGCAGGAGGCATGAGCCCTTCCCT
Phenotype
MIM: 607114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM33 PubMed Links

Gene Details

Gene
ADAM33
Gene Name
ADAM metallopeptidase domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282447.2 Intron NP_001269376.1
NM_025220.4 Intron NP_079496.1
NM_153202.3 Intron NP_694882.1
XM_005260843.1 Intron XP_005260900.1
XM_006723639.1 Intron XP_006723702.1
XM_006723640.1 Intron XP_006723703.1
XM_006723644.2 Intron XP_006723707.1
XM_011529366.1 Intron XP_011527668.1
XM_011529367.1 Intron XP_011527669.1
XM_011529368.1 Intron XP_011527670.1
XM_011529370.1 Intron XP_011527672.1
XM_011529371.1 Intron XP_011527673.1
XM_011529373.1 Intron XP_011527675.1
XM_017028080.1 Intron XP_016883569.1
XM_017028081.1 Intron XP_016883570.1
XM_017028082.1 Intron XP_016883571.1
XM_017028083.1 Intron XP_016883572.1
Gene
GFRA4
Gene Name
GDNF family receptor alpha 4
There are no transcripts associated with this gene.

View Full Product Details