Product Details

SNP ID
rs2280169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108290853 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCCCAAAGCTGCATTTTGGACC[C/T]GTCTCCAAAGGCGTTGAGGAAATTA
Phenotype
MIM: 602351
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMKLR1 PubMed Links

Gene Details

Gene
CMKLR1
Gene Name
chemerin chemokine-like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142343.1 2687 UTR 3 NP_001135815.1
NM_001142344.1 2687 UTR 3 NP_001135816.1
NM_001142345.1 2687 UTR 3 NP_001135817.1
NM_004072.2 2687 UTR 3 NP_004063.1
XM_017018820.1 2687 UTR 3 XP_016874309.1

View Full Product Details