Product Details
- SNP ID
-
rs2293368
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:185370476 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATTATGCTAATAAGATTCTGACAG[A/G]ATTGTCTGCAAACAATCTGCATGTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LRP2BP
PubMed Links
Gene Details
- Gene
- LRP2BP
- Gene Name
- LRP2 binding protein
- Gene
- SNX25
- Gene Name
- sorting nexin 25
There are no transcripts associated with this gene.
View Full Product Details