Product Details

SNP ID
rs2293448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48924069 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACAGCGCAAGAGGCACAGAGGCT[C/T]TGAATGCCCTCAGAGGTCCCACTGA
Phenotype
MIM: 103190 MIM: 611088 MIM: 610571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARF3 PubMed Links

Gene Details

Gene
ARF3
Gene Name
ADP ribosylation factor 3
There are no transcripts associated with this gene.

Gene
CCDC65
Gene Name
coiled-coil domain containing 65
There are no transcripts associated with this gene.

Gene
FKBP11
Gene Name
FK506 binding protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143781.1 Intron NP_001137253.1
NM_001143782.1 Intron NP_001137254.1
NM_016594.2 Intron NP_057678.1

View Full Product Details