Product Details

SNP ID
rs2303482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48578164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACGCCGCGCGCCGCCCGAAGCC[A/C]GCCTCCGGCTGGAAGCTGCTCTCTC
Phenotype
MIM: 142966 MIM: 142965 MIM: 610173
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOXB3 PubMed Links

Gene Details

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 218 Intron NP_002137.4
XM_005257277.3 218 Intron XP_005257334.1
XM_005257280.3 218 Intron XP_005257337.1
XM_005257282.3 218 Intron XP_005257339.1
XM_006721854.2 218 Intron XP_006721917.1
XM_011524707.2 218 Intron XP_011523009.1
XM_011524708.2 218 Intron XP_011523010.1
XM_011524710.1 218 Intron XP_011523012.1
XM_011524719.1 218 Intron XP_011523021.1
XM_011524720.2 218 Intron XP_011523022.1
XM_011524721.2 218 Intron XP_011523023.1
XM_011524726.2 218 Intron XP_011523028.1
XM_017024557.1 218 Intron XP_016880046.1
XM_017024558.1 218 Intron XP_016880047.1
XM_017024559.1 218 Intron XP_016880048.1
XM_017024560.1 218 Intron XP_016880049.1
XM_017024561.1 218 Intron XP_016880050.1
XM_017024562.1 218 Intron XP_016880051.1
XM_017024563.1 218 Intron XP_016880052.1
Gene
HOXB4
Gene Name
homeobox B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024015.4 218 Silent Mutation GCG,GCT A52A NP_076920.1
Gene
MIR10A
Gene Name
microRNA 10a
There are no transcripts associated with this gene.

View Full Product Details