Product Details

SNP ID
rs2099099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34368552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATGGCTGCCTGGGGTTGGGGGGG[A/G]CAGTCTTTATATCCTGACCGTAATC
Phenotype
MIM: 172400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPI PubMed Links

Gene Details

Gene
GPI
Gene Name
glucose-6-phosphate isomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184722.1 Intron NP_001171651.1
NM_001289789.1 Intron NP_001276718.1
XM_005258764.2 Intron XP_005258821.1
XM_006723148.2 Intron XP_006723211.1
XM_011526754.2 Intron XP_011525056.1

View Full Product Details