Product Details

SNP ID
rs2304491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70382792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGGGTTCCCACCACCCACACCA[T/C]GGGCCTACCCACCTCATCACACACA
Phenotype
MIM: 607188
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DDX19A PubMed Links

Gene Details

Gene
DDX19A
Gene Name
DEAD-box helicase 19A
There are no transcripts associated with this gene.

Gene
ST3GAL2
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006927.3 Intron NP_008858.1

View Full Product Details