Product Details

SNP ID
rs2304692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45081105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGATCCCCCATTCCACCACAGC[C/T]GCCACCCAGACCTTGGCCTCGCCAA
Phenotype
MIM: 607929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCM2 PubMed Links

Gene Details

Gene
CCM2
Gene Name
CCM2 scaffolding protein
There are no transcripts associated with this gene.

Gene
LOC100128364
Gene Name
uncharacterized LOC100128364
There are no transcripts associated with this gene.

Gene
NACAD
Gene Name
NAC alpha domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146334.1 Intron NP_001139806.1
XM_006715674.3 Intron XP_006715737.1

View Full Product Details