Product Details
- SNP ID
-
rs2132428
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:212994420 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAGCTTTACACGTTATGCTGGCTT[A/G]TATTTCCCAAACATTTCCCCAACAC
- Phenotype
-
MIM: 610471
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANGEL2
PubMed Links
Gene Details
- Gene
- ANGEL2
- Gene Name
- angel homolog 2
- Gene
- VASH2
- Gene Name
- vasohibin 2
There are no transcripts associated with this gene.
View Full Product Details