Product Details

SNP ID
rs3024904
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:191030476 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGCCATTGACTGTTATTCATATAT[A/T]ATTGATGAGGCTCCGGGTTCTAGAA
Phenotype
MIM: 600558
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105373805 PubMed Links

Gene Details

Gene
LOC105373805
Gene Name
uncharacterized LOC105373805
There are no transcripts associated with this gene.

Gene
STAT4
Gene Name
signal transducer and activator of transcription 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243835.1 Intron NP_001230764.1
NM_003151.3 Intron NP_003142.1
XM_006712719.3 Intron XP_006712782.1
XM_011511705.2 Intron XP_011510007.1
XM_017004784.1 Intron XP_016860273.1

View Full Product Details