Product Details

SNP ID
rs2347685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134649639 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACAAAACCCTTCTGCTGAAACTT[A/G]GCCACAAATATGAGGAAAACATACT
Phenotype
MIM: 613896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPGM PubMed Links

Gene Details

Gene
BPGM
Gene Name
bisphosphoglycerate mutase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293085.1 Intron NP_001280014.1
NM_001724.4 Intron NP_001715.1
NM_199186.2 Intron NP_954655.1
XM_011516527.1 Intron XP_011514829.1

View Full Product Details