Product Details

SNP ID
rs2365716
Assay Type
Functionally tested
NCBI dbSNP Submissions
66
Location
Chr.1:156645402 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGAATGTTGGGGAGTAAAGAGA[A/G]GAGTTTCCTTACAAATAATGGTGGG
Phenotype
MIM: 600347
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAN PubMed Links

Gene Details

Gene
BCAN
Gene Name
brevican
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021948.4 142 Intron NP_068767.3
NM_198427.1 142 Intron NP_940819.1
XM_011509866.1 142 Intron XP_011508168.1
XM_017002047.1 142 Missense Mutation AAG,AGG K7R XP_016857536.1

View Full Product Details