Product Details

SNP ID
rs2391265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26204745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACTTGTGGGATGGCTTTAATGA[C/T]TGAGCAATTAAGGTTTATCTTTTTT
Phenotype
MIM: 604477 MIM: 600124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBX3 PubMed Links

Gene Details

Gene
CBX3
Gene Name
chromobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007276.4 Intron NP_009207.2
NM_016587.3 Intron NP_057671.2
XM_005249611.3 Intron XP_005249668.1
Gene
HNRNPA2B1
Gene Name
heterogeneous nuclear ribonucleoprotein A2/B1
There are no transcripts associated with this gene.

View Full Product Details