Product Details

SNP ID
rs2453645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100158185 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCGCATCTCAGCCCTTCCCTCCC[A/G]GAGCGGGGGCGAGAGGTCGAGGCGG
Phenotype
MIM: 609110 MIM: 606033 MIM: 603395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO43 PubMed Links

Gene Details

Gene
FBXO43
Gene Name
F-box protein 43
There are no transcripts associated with this gene.

Gene
POLR2K
Gene Name
RNA polymerase II subunit K
There are no transcripts associated with this gene.

Gene
SPAG1
Gene Name
sperm associated antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003114.4 Intron NP_003105.2
NM_172218.2 Intron NP_757367.1
XM_011517240.2 Intron XP_011515542.1
XM_011517241.2 Intron XP_011515543.1
XM_011517242.2 Intron XP_011515544.1
XM_011517243.2 Intron XP_011515545.1
XM_011517245.2 Intron XP_011515547.1
XM_017013754.1 Intron XP_016869243.1
XM_017013755.1 Intron XP_016869244.1

View Full Product Details