Product Details

SNP ID
rs2435611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151178911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGCCCGTCTCTGTCCTTCCTGG[A/C]CCATGTTGATCCCCACCTCTGCACT
Phenotype
MIM: 615054
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASB10 PubMed Links

Gene Details

Gene
ASB10
Gene Name
ankyrin repeat and SOCS box containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142459.1 Intron NP_001135931.2
NM_001142460.1 Intron NP_001135932.2
NM_080871.3 Intron NP_543147.2

View Full Product Details