Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001034836.1 | Intron | NP_001030008.1 | |||
NM_001163120.1 | Intron | NP_001156592.1 | |||
NM_001163121.1 | Intron | NP_001156593.1 | |||
NM_001163122.1 | Intron | NP_001156594.1 | |||
NM_001163124.1 | Intron | NP_001156596.1 | |||
NM_001163125.1 | Intron | NP_001156597.1 | |||
NM_001163130.1 | Intron | NP_001156602.1 | |||
NM_145654.3 | Intron | NP_663629.1 | |||
XM_011524509.2 | Intron | XP_011522811.1 | |||
XM_011524510.2 | Intron | XP_011522812.1 |